Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDGF-BB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588272 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588272 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588272 Supplier Invitrogen™ Supplier No. PA588272
  • $588.00
    $484.65 / Each of 1

    Save $103.35

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Positive Samples: SW480, MCF7, Mouse skeletal muscle Immunogen sequence: QCRPTQVQLR PVQVRKIEIV RKKPIFKKAT VTLEDHLACK CETVAAARPV TRSPGGSQEQ RAKTPQTRVT IRTVRVRRPP KGKHRKFKHT HDKTALKETL GA.

The protein encoded by this gene is a member of the platelet-derived growth factor family. The four members of this family are mitogenic factors for cells of mesenchymal origin and are characterized by a motif of eight cysteines. This gene product can exist either as a homodimer (PDGF-BB) or as a heterodimer with the platelet-derived growth factor alpha polypeptide (PDGF-AB), where the dimers are connected by disulfide bonds. Mutations in this gene are associated with meningioma. Reciprocal translocations between chromosomes 22 and 7, at sites where this gene and that for COL1A1 are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans resulting from unregulated expression of growth factor. Two alternatively spliced transcript variants encoding different isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDGF-BB
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.58 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene PDGFB
Gene Accession No. P01127, Q05028
Gene Alias becaplermin; c-sis; FLJ12858; H-PDGF-BB; IBGC5; PDGF; pdgf protein; PDGF subunit B; PDGF, B chain; PDGF2; PDGF-2; PDGFB; PDGF-B; platelet derived growth factor subunit B; platelet derived growth factor, B polypeptide; platelet-derived growth factor 2; platelet-derived growth factor B; platelet-derived growth factor B chain; platelet-derived growth factor beta; platelet-derived growth factor beta polypeptide; platelet-derived growth factor beta polypeptide (simian sarcoma viral (v-sis) oncogene homolog); platelet-derived growth factor subunit B; platelet-derived growth factor, beta polypeptide (oncogene SIS); Platelet-derived growth factor, same as Sis (Simian sarcoma viral oncogene homologue, c-sis); proto-oncogene c-Sis; Sis; SSV
Gene Symbols PDGFB
Host Species Rabbit
Immunogen Synthetic peptide.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24628, 5155
Target Species Human, Rat, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.