Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ PDGFA Protein
SDP

Catalog No. NBP14314803 Shop All R&D Systems Products
Change view
Click to view available options
0.025mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
NBP14314803 0.025mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP14314803 Supplier Novus Biologicals™ Supplier No. NBP1431480.025MG
Add to Cart
Add to Cart

Highly purified and high bioactivity. Generating reliable and reproducible results.

Recombinant non-glycosylated homodimer of 126 amino acids corresponding to PDGF. Source: E. coli The Recombinant Mouse PDGFA Protein is derived from E. coli. The Recombinant Mouse PDGFA Protein has been validated for the following applications: Immunoassay, Bioactivity.

Specifications

Formulation phosphate buffered saline, 1% BSA.
Quantity 0.025mg
Source Human
Storage Requirements Store at -80°C. Avoid freeze-thaw cycles.
Common Name PDGFA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.