Learn More
Enzo Life Sciences PDGF receptor beta monoclonal antibody (42G12) (100µg)
SDP
Supplier:  Enzo Life Sciences ADI905682100
Encompass_Preferred
Clone: 42G12. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant human PDGF Receptor β. UniProt ID: P09619. Source: Purified from mouse ascites. Species reactivity: Mouse, Rat. Creossreactivity: Weakly cross-reacts with human PDGF receptor β. Applications: IHC (PS), WB. Recommended Dilutions: Western Blot (1µg/ml, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 0.09% sodium azide and 50% glycerol. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-6990
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.