Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDI Monoclonal Antibody (RL90), Alexa Fluor™ 555
GREENER_CHOICE

Catalog No. PIMA3019AZ Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA3019AZ 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA3019AZ Supplier Invitrogen™ Supplier No. MA3019A555
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

MA3019A555 detects protein disulphide-isomerase (PDI) from human, rat, porcine and mouse tissues as well as hamster cells. The MA3019A555 antigen is purified rat PDI.

Protein disulfide isomerase (PDI) is a multifunctional protein that catalyzes the formation, breakage and rearrangement of disulfide bonds. At the cell surface, the protein seems to act as a reductase that cleaves disulfide bonds of proteins attached to the cell. It may therefore cause structural modifications of exofacial proteins. Inside the cell, PDI seems to form/rearrange disulfide bonds of nascent proteins. At high concentrations, it functions as a chaperone that inhibits aggregation of misfolded proteins. At low concentrations, it facilitates aggregation (anti-chaperone activity). PDI may be involved with other chaperones in the structural modification of the TG precursor in hormone biogenesis. It also acts a structural subunit of various enzymes such as prolyl 4-hydroxylase and microsomal triacylglycerol transfer protein MTTP. Receptor for LGALS9; the interaction retains P4HB at the cell surface of Th2 T helper cells, increasing disulfide reductase activity at the plasma membrane, altering the plasma membrane redox state and enhancing cell migration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDI
Applications Immunocytochemistry
Classification Monoclonal
Clone RL90
Concentration 1 mg/mL
Conjugate Alexa Fluor 555
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
Gene P4hb
Gene Accession No. P04785, P07237, P09103, Q8R4U2
Gene Alias 1810041F13Rik; AI661267; CaO19.12595; cellular thyroid hormone binding protein; cellular thyroid hormone-binding protein; CLCRP1; collagen prolyl 4-hydroxylase beta; DSI; Endoplasmic reticulum resident protein 59; Eps1p; ER protein 59; ERBA2L; ERp59; GIT; glutathione-insulin transhydrogenase; I79_002399; LOW QUALITY PROTEIN: protein disulfide-isomerase A2; MFP1; P4HB; p4hb.L; P4Hbeta; p55; p58; Pancreas-specific protein disulfide isomerase; pancreatic protein disulfide isomerase; PDA2; PDI; PDI (E.C.5.3.4.1); PDI1; PDI1P; Pdia1; pdia2; pdia2.L; PDIP; Pdipl; PDIR; PHDB; PO4DB; PO4HB; procollagen-proline, 2-oxoglutarate 4-dioxygenase (proline 4-hydroxylase), beta polypeptide; PROHB; prolyl 4-hydroxylase beta polypeptide; Prolyl 4-hydroxylase subunit beta; prolyl 4-hydroxylase subunit beta L homeolog; prolyl 4-hydroxylase subunit beta L homeolog; prolyl 4-hydroxylase, beta polypeptide L homeolog; prolyl 4-hydroxylase, beta polypeptide; prolyl 4-hydroxylase, beta subunit; protein disulfide isomerase; protein disulfide isomerase (pancreas) like; Protein disulfide isomerase (Prolyl 4-hydroxylase, beta polypeptide); protein disulfide isomerase A2; protein disulfide isomerase associated 2; protein disulfide isomerase family A member 2; protein disulfide isomerase family A member 2 L homeolog; protein disulfide isomerase family A, member 1; protein disulfide isomerase family A, member 2; protein disulfide isomerase PDI1; protein disulfide isomerase, pancreatic; protein disulfide isomerase/oxidoreductase; protein disulfide isomerase-associated 1; protein disulfide isomerase-associated 2; protein disulfide-isomerase; Protein disulfide-isomerase A2; protocollagen hydroxylase; Rho GDP dissociation inhibitor gamma; testicular secretory protein Li 32; Thbp; Thioredoxin-related glycoprotein 1; thyroid hormone-binding protein p55; TRG1; XELAEV_18043753mg; XELAEV_18045582mg; XPDIp; YCL043C; YCL313; YCL43C
Gene Symbols P4hb
Host Species Mouse
Immunogen Purified rat PDI protein.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100689433, 110255932, 18453, 25506, 5034
Target Species Hamster, Human, Mouse, Pig, Rat
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.