Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDIA6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA3008 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA3008 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA3008 Supplier Invitrogen™ Supplier No. PA3008
  • $646.33
    $502.65 / Each of 1

    Save $143.68

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

PA3-008 detects P5 protein in human samples. PA3-008 has successfully been used in Western blot procedures. By Western blot, this antibody detects an ~50 kDa protein representing P5 in human fibrosarcoma HT 1080 cell lysate. The PA3-008 immunogen is a synthetic peptide corresponding to residues D(476) I D L S D V E L D D L G K D E L(492) of human P5. This peptide (Cat. # PEP-224) is available for use in neutralization and control experiments.

P5 is a novel protein that was originally identified in humans. Functionally, P5 has been shown to have peptide binding ability and chaperone activity specific to certain proteins. P5 shares several structural similarities to PDI, an important protein of the ER. The deduced amino acid sequence of P5 shows that it contains the ER retention signal KDEL. P5 also contains two CXXC-like motifs, a motif responsible for oxidoreductase activity. These two CXXC motifs are found on the N- and C-terminus of the protein, one at each end. Studies suggest that the N-terminal CXXC motif is more important than the second for isomerase activity. Contrary to most of the PDI-like proteins, P5 is not stress-inducible.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDIA6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 0.01% sodium azide
Gene PDIA6
Gene Accession No. Q15084
Gene Alias 1700015E05Rik; AL023058; C77895; CaBP1; CaBP5; Calcium-binding protein 1; endoplasmic reticulum protein 5; ER protein 5; ERP5; P5; PDIA6; pdi-p5; protein disulfide isomerase A6; protein disulfide isomerase associated 6; protein disulfide isomerase family A member 6; protein disulfide isomerase family A, member 6; protein disulfide isomerase P5; protein disulfide isomerase-associated 6; protein disulfide isomerase-related protein; protein disulfide-isomerase A6; thioredoxin domain containing 7; thioredoxin domain containing 7 (protein disulfide isomerase); thioredoxin domain-containing protein 7; TXNDC7
Gene Symbols PDIA6
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues D(476) I D L S D V E L D D L G K D E L(492) of human P5.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10130
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.