Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDIK1L Monoclonal Antibody (4B7)
GREENER_CHOICE

Catalog No. PIMA523088
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA523088 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA523088 Supplier Invitrogen™ Supplier No. MA523088
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: MVSSQPKYDL IREVGRGSYG VVYEAVIRKT SARVAVKKIR CHAPENVELA LREFWALSSI KSQHPNVIHL EECILQKDGM VQKMSHGSNS SLYLQLVET.

PDIK1L is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDIK1L
Applications ELISA
Classification Monoclonal
Clone 4B7
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PDIK1L
Gene Accession No. Q8N165
Gene Alias BC027088; casein kinase; CLIK1L; PDIK1L; PDLIM1 interacting kinase 1 like; PDLIM1-interacting kinase 1-like; Serine/threonine-protein kinase PDIK1L; STK35L2
Gene Symbols PDIK1L
Host Species Mouse
Immunogen PDIK1L (NP_690048, 1 a.a. ~ 99 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 149420
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.