Learn More
AdipoGen PDITC
SDP
Supplier:  AdipoGen CDXP0022G020
Encompass_Preferred
Chemical. CAS 4044-65-9. Formula C8H4N2S2. MW 192.26. Synthetic. Reagent for solid-phase sequencing.
Catalog No. 50-256-0273
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.