Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDK3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528019 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528019 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528019 Supplier Invitrogen™ Supplier No. PA528019
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2, mouse liver, PDK3-transfected 293T. Predicted reactivity: Mouse (96%), Rat (96%), Chicken (93%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PDK3 inhibits the mitochondrial pyruvate dehydrogenase complex by phosphorylation of the E1 alpha subunit, thus contributing to the regulation of glucose metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDK3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.16 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene PDK3
Gene Accession No. Q15120, Q922H2
Gene Alias [Pyruvate dehydrogenase (acetyl-transferring)] kinase isozyme 3, mitochondrial; [Pyruvate dehydrogenase [lipoamide]] kinase isozyme 3, mitochondrial; 2610001M10Rik; AI035637; CMTX6; GS1-358P8.4; PDHK3; PDK3; pyruvate dehydrogenase; Pyruvate dehydrogenase kinase 3; Pyruvate dehydrogenase kinase isoform 3; pyruvate dehydrogenase kinase, isoenzyme 3; pyruvate dehydrogenase kinase, isozyme 3; pyruvate dehydrogenase, lipoamide, kinase isozyme 3, mitochondrial
Gene Symbols PDK3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PDK3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 236900, 5165
Target Species Amphibian, Human, Mouse, Rodent
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.