Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDLIM5 Monoclonal Antibody (OTI1A11), TrueMAB™, OriGene
SDP

Catalog No. 502540317
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0317 30 μL
50-167-5139 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0317 Supplier OriGene Supplier No. TA504449S
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PDLIM5 Monoclonal antibody specifically detects PDLIM5 in Human samples. It is validated for Flow Cytometry, Western Blot

Specifications

Antigen PDLIM5
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone OTI1A11
Concentration 3.37 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PDLIM5
Gene Accession No. Q96HC4
Gene Alias 1110001A05Rik; AI987914; C87059; ENH; Enh1; Enh2; Enh3; Enigma homolog; enigma-like LIM domain protein; enigma-like PDZ and LIM domains protein; L9; LIM; Pdlim5; PDZ and LIM domain 5; PDZ and LIM domain protein 5
Host Species Mouse
Immunogen Full length human recombinant protein of human PDLIM5 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10611
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.