Learn More
Cayman Chemical PDM 2 Fatty Acids and Derivati
SDP
Supplier:  Cayman Chemical 1000634210 MG
Encompass_Preferred
A resveratrol-based AhR antagonist (Ki = 1.2 nM); selectively binds to the AhR over the estrogen receptor at 100 µM
Catalog No. 50-288-8160