Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PDRG Polyclonal Antibody, QED Bioscience™
SDP

Catalog No. 502277977
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-7977 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-7977 Supplier Leinco Technologies Inc Supplier No. 34012100UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PDRG Polyclonal antibody specifically detects PDRG in Human samples. It is validated for Western Blot

Specifications

Antigen PDRG
Applications Western Blot
Classification Polyclonal
Concentration 0.5-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PDRG1
Gene Accession No. Q9NUG6
Gene Alias 1110004D19Rik; AU020945; AV006589; C20orf126; p53 and DNA damage regulated 1; p53 and DNA damage-regulated protein 1; p53 and DNA-damage regulated 1; PDRG; PDRG1; RGD1305096
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the aa sequence (16-96) CDHLDKEIEKLRKQLK of human PDRG conjugated to KLH.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81572
Target Species Human
Content And Storage 2-8°C
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.