Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PDSS2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521626
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521626 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521626 Supplier Invitrogen™ Supplier No. PA521626
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431, mouse brain. Predicted reactivity: Mouse (89%), Rat (87%), Pig (96%), Rhesus Monkey (98%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The isoprenoid chain of ubiquinone (coenzyme Q) varies in length between species and is determined by trans-polyprenyl diphosphate synthase. Humans possess a heterotetrameric decaprenyl diphosphate synthase composed of DPS1 and DLP1 (PDSS2) that produces Q10 ubiquinone.The isoprenoid chain of ubiquinone (coenzyme Q) varies in length between species and is determined by trans-polyprenyl diphosphate synthase. Humans possess a heterotetrameric decaprenyl diphosphate synthase composed of DPS1 and DLP1 (PDSS2) that produces Q10 ubiquinone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PDSS2
Applications Western Blot
Classification Polyclonal
Concentration 0.54 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene PDSS2
Gene Accession No. Q33DR3, Q86YH6
Gene Alias 5430420P03Rik; All-trans-decaprenyl-diphosphate synthase subunit 2; bA59I9.3; C6orf210; Candidate tumor suppressor protein; COQ10D3; decaprenyl diphosphate synthase subunit 2; decaprenyl pyrophosphate synthase subunit 2; decaprenyl pyrophosphate synthetase subunit 2; decaprenyl-diphosphate synthase subunit 2; DLP1; EGK_15212; Gm60; hDLP1; hypothetical protein LOC436624; kd; mDLP1; Pdss2; Plmp; prenyl (decaprenyl) diphosphate synthase, subunit 2; prenyl (solanesyl) diphosphate synthase, subunit 2; prenyl diphosphate synthase, subunit 2; prenyltransferase-like mitochondrial protein; RP11-59I9.3; solanesyl-diphosphate synthase subunit 2; subunit 2 of decaprenyl diphosphate synthase; zgc:92156
Gene Symbols PDSS2
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PDSS2. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57107, 71365
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.