Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ PE 22-28
GREENER_CHOICESDP

Catalog No. 78681
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
78-681 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 78-681 Supplier Tocris Bioscience™ Supplier No. 7868/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent inhibitor of TREK-1 channels

PE 22-28 is a potent inhibitor of potassium channel subfamily K member 2/TREK-1 (IC50 = 0.12 nM). In rodent models of ischemia PE 22-28 reduces apoptotic death of β cells, increases hippocampal neurogenesis, and reduces stroke-induced motor and cognitive defects.

Specifications

Purity 95%
CAS 1801959-12-5
Content And Storage Store at -20°C
Quantity 1 mg
Target Entry TREK 1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.