Learn More
Biolegend PE anti-human CD253 (Trail)
Supplier:  Biolegend 308206
Encompass_Preferred
PE anti-human CD253 (Trail) [RIK-2]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green; Apps: FC; Size: 100 tests
Catalog No. 50-166-469