Learn More
Biolegend PE anti-human IgM
Supplier:  Biolegend 314507
Encompass_Preferred
PE anti-human IgM [MHM-88]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC, ICFC; Size: 25 tests
Catalog No. 50-166-808