Learn More
ABclonal Technology PE anti-Human CD170/Siglec-5
SDP
Supplier:  ABclonal Technology A26119100T
Encompass_Preferred
This gene encodes a member of the sialic acid-binding immunoglobulin-like lectin (Siglec) family These cell surface lectins are characterized by structural motifs in the immunoglobulin (Ig)-like domains and sialic acid recognition sites in the first Ig V set domain The encoded protein is a member of the CD33-related subset of Siglecs and inhibits the activation of several cell types including monocytes macrophages and neutrophils Binding of group B Streptococcus (GBS) to the encoded protein plays a role in GBS immune evasion
Catalog No. 50-000-09632
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.