Learn More
ABclonal Technology PE anti-Human ICOS/CD278
SDP
Supplier:  ABclonal Technology A26444500T
Encompass_Preferred
The protein encoded by this gene belongs to the CD28 and CTLA-4 cell-surface receptor family It forms homodimers and plays an important role in cell-cell signaling immune responses and regulation of cell proliferation
Catalog No. 50-000-10776
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.