Learn More
Cytek Biosciences Inc Antibodies PE Anti-Mouse CD80 (B7-1) (16-10A1) ..... Cat. No. 50-0801-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 500801U025
Encompass_Preferred
Antibodies PE Anti-Mouse CD80 (B7-1) (16-10A1) ..... Cat. No. 50-0801-U025 ..... Size 25 µgThe 16-10A1 antibody reacts with mouse CD80, also known as B7-1, a 55 kDa type I transmembrane protein ligand for CD152 (CTLA-4) and for CD28, a co-stimulatory receptor for the T cell receptor (TCR). CD28 also binds a second B7 ligand known as CD86 (B7-2). Both CD80 and CD86 are expressed on activated B cells and antigen-presenting cells. These ligands trigger CD28 signaling in concert with TCR activation to drive T cell proliferation, induce high-level expression of IL-2, impart resistance to apoptosis, and enhance T cell cytotoxicity. The interaction / co-stimulatory signaling between the B7 ligands and CD28 or CTLA-4 provides crucial communication between T cells and B cells or APCs to coordinate the adaptive immune response.
Catalog No. 50-210-3411
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.