Learn More
Cytek Biosciences Inc Antibodies PE Anti-Mouse CD86 (B7-2) (PO3.1) ..... Cat. No. 50-0861-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 500861U025
Encompass_Preferred
Antibodies PE Anti-Mouse CD86 (B7-2) (PO3.1) ..... Cat. No. 50-0861-U025 ..... Size 25 µgThe PO3.1 antibody reacts with mouse CD86, also known as B7-2, an 80 kDa cell surface protein which is a ligand for CD28, a co-stimulatory receptor for the T cell receptor (TCR). CD28 can also bind a second B7 ligand known as CD80 (B7-1). Both CD80 and CD86 are expressed on activated B cells and antigen-presenting cells. These ligands trigger CD28 signaling in concert with TCR activation to drive T cell proliferation, induce high-level expression of IL-2, impart resistance to apoptosis, and enhance T cell cytotoxicity. The interaction / co-stimulatory signaling between the B7 ligands and CD28 provides crucial communication between T cells and B cells or APCs to coordinate the adaptive immune response.The PO3.1 antibody may be used as a marker for CD86 expression on B cells, macrophages, and dendritic cells.
Catalog No. 50-210-3413
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.