Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PE-CF594 Mouse Anti-Human FoxJ2, BD Horizon™

Catalog No. BDB571076 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB571076 100 Tests
BDB571079 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB571076 Supplier BD Biosciences Supplier No. 571076
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

FoxJ2 Monoclonal Antibody Specifically detects FoxJ2 in Human Samples. It is validated for Intracellular Staining (Flow Cytometry).
TRUSTED_SUSTAINABILITY

Specifications

Antigen FoxJ2
Applications Flow Cytometry (Intracellular Staining)
Classification Monoclonal
Clone N17-280
Conjugate PE-CF594
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias Forkhead box J2, HNF3B, HNF-3B, HNF-3β, hepatocyte nuclear factor 3β
Host Species Mouse
Immunogen Human FoxJ2 Peptide
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3170
Target Species Human
Content And Storage The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.