Learn More
Biolegend PE/Cyanine5 anti-human CD29
Supplier:  Biolegend 303006
Encompass_Preferred
PE/Cyanine5 anti-human CD29 [TS2/16]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cattle (Bovine, Cow), Cynomolgus, Dog (Canine), (Horse reactivity), Rhesus; Apps: FC; Size: 100 tests
Catalog No. 50-165-941
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.