Learn More
Biolegend PE/Cyanine7 anti-human HLA-DR, DP, DQ
Supplier:  Biolegend 361708
Encompass_Preferred
PE/Cyanine7 anti-human HLA-DR, DP, DQ [Tü39]; Isotype: Mouse IgG2a, κ; Reactivity: Human, Cross-Reactivity: Baboon, Cynomolgus, Cat (Feline), Dog (Canine), Cattle; Apps: FC; Size: 100 tests
Catalog No. 50-103-0811
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.