Learn More
Biolegend PE/Cyanine7 anti-mouse H-2Kb Antibody, 100 ug
Supplier:  Biolegend 116520
Encompass_Preferred
The AF6-88.5 antibody reacts with H-2Kb MHC class I alloantigen expressed on nucleated cells from mice of the H-2Kb haplotype. H-2Kb is involved in antigen presentation to T cells expressing CD3/TCR and CD8 proteins. The AF6-88.5 antibody does not cross-react with other haplotypes (d, f, j, k, p, q, r, s, u, v).
Catalog No. NC1613943
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.