Learn More
Cytek Biosciences Inc PE-CY7 MS CD11C (N418) 100UG
Supplier:  Cytek Biosciences Inc 600114U100
Encompass_Preferred
PE-Cyanine7 Anti-Mouse CD11c (N418) Isotype: Armenian Hamster IgG Apps: FC Size: 100 ug
Catalog No. 50-105-4846
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.