Learn More
Biolegend PE/Cyanine7 anti-mouse GARP (LRRC32)
Supplier:  Biolegend 142910
Encompass_Preferred
PE/Cyanine7 anti-mouse GARP (LRRC32) [F011-5]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1692008