Learn More
Cytek Biosciences Inc Antibodies PE-Cyanine7 Anti-Human CD45RA (HI100) ..... Cat. No. 60-0458-T025 ..... Size 25 tests
Supplier:  Cytek Biosciences Inc 600458T025
Encompass_Preferred
Antibodies PE-Cyanine7 Anti-Human CD45RA (HI100) ..... Cat. No. 60-0458-T025 ..... Size 25 testsThe HI100 antibody reacts with the human CD45 isoform known as CD45RA, a protein tyrosine phosphatase of ‰¥ 220 kDa. CD45 is one of the most abundant hematopoietic markers, and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). Various isoforms are generated and expressed in cell-specific patterns. With their broad cell distribution, CD45 isoforms are critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RB and CD45RO.The HI100 antibody is widely used as a marker for human CD45RA expression on naive and activated T cells, B cells, and monocytes.
Catalog No. 50-210-3524
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.