Learn More
Cytek Biosciences Inc Antibodies PE-Cyanine7 Anti-Human CD69 (FN50) ..... Cat. No. 60-0699-T025 ..... Size 25 tests
Supplier:  Cytek Biosciences Inc 600699T025
Encompass_Preferred
Antibodies PE-Cyanine7 Anti-Human CD69 (FN50) ..... Cat. No. 60-0699-T025 ..... Size 25 testsThe FN50 monoclonal antibody reacts with human CD69, a type II transmembrane glycoprotein also known as the Very Early Activation Antigen, EA-1, Leu23, Activation Inducer Molecule (AIM) and CLEC2C. CD69 is expressed as a disulfide-linked homodimer on activated T and B cells, NK cells, neutrophils, and monocytes. Induction occurs rapidly upon activation and is transient. It is also constitutively expressed on platelets and a subset of thymocytes.
Catalog No. 50-210-3530
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.