Learn More
Cytek Biosciences Inc Antibodies PE-Cyanine7 Anti-Mouse CD16 / CD32 (2.4G2) ..... Cat. No. 60-0161-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 600161U025
Encompass_Preferred
Antibodies PE-Cyanine7 Anti-Mouse CD16 / CD32 (2.4G2) ..... Cat. No. 60-0161-U025 ..... Size 25 µgThe 2.4G2 antibody is specific for a common epitope found in the extracellular regions of mouse Fc-receptors Fc-gamma II (CD32) and Fc-gamma III (CD16). These receptors are expressed by B cells, monocytes, macrophages, NK cells, dendritic cells, and neutrophils.
Catalog No. 50-210-3507
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.