Learn More
Cytek Biosciences Inc Antibodies PE-Cyanine7 Anti-Mouse CD45 (30-F11) ..... Cat. No. 60-0451-U025 ..... Size 25 µg
Supplier:  Cytek Biosciences Inc 600451U025
Encompass_Preferred
Antibodies PE-Cyanine7 Anti-Mouse CD45 (30-F11) ..... Cat. No. 60-0451-U025 ..... Size 25 µgThe 30-F11 antibody reacts with mouse CD45, which is one of the most abundant hematopoietic markers and is expressed on all leukocytes (the Leukocyte Common Antigen, LCA). CD45 is a protein tyrosine phosphatase existing in several isoforms, each being generated and expressed in cell-specific patterns. With its broad cell distribution, CD45 is critical for many leukocyte functions, regulating signal transduction and cell activation associated with the T cell receptor, B cell receptor, and IL-2 receptor. Other forms of CD45, with restricted cellular expression, include CD45R (B220), CD45RA, CD45RB, CD45RO and others.The 30-F11 antibody is widely used as a leukocyte marker for B cells, T cell subsets and NK cell subsets.
Catalog No. 50-210-3520
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.