Learn More
Biolegend PE/Dazzle™ 594 anti-mouse VISTA (PD-1H)
Supplier:  Biolegend 150214
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse VISTA (PD-1H) [MIH63]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 ug
Catalog No. NC1540269