Learn More
Biolegend PE/DazzleTM 594 anti-mouse CD2
Supplier:  Biolegend 123820
Encompass_Preferred
PE/Dazzle™ 594 anti-mouse CD200 (OX2) [OX-90]; Isotype: Rat IgG2a, κ; Reactivity: Mouse; Apps: FC; Size: 100 μg
Catalog No. 50-272-2719
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.