Learn More
Biolegend PE/DazzleTM 594 anti-rat CD90/
Supplier:  Biolegend 202541
Encompass_Preferred
PE/Dazzle™ 594 anti-rat CD90/mouse CD90.1 (Thy-1.1) [OX-7]; Isotype: Mouse IgG1, κ; Reactivity: Rat, Mouse; Apps: FC; Size: 25 μg
Catalog No. 50-272-2551