Learn More
Biolegend PE/Dazzle™ 594 anti-human IFN-γ
Supplier:  Biolegend 506530
Encompass_Preferred
PE/Dazzle™ 594 anti-human IFN-γ [B27]; Isotype: Mouse IgG1, κ; Reactivity: Human, Cross-Reactivity: Chimpanzee, Baboon, Cynomolgus, Rhesus, African Green Monkey, Pigtailed Macaque, Sooty Mangabey; Apps: ICFC; Size: 100 tests
Catalog No. 50-208-5762
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.