Learn More
ACROBiosystems PE-Labeled Human HLA-A0201B
Supplier:  ACROBiosystems HLHHP2H6200UG
Encompass_Preferred
PE-Labeled Human HLA-A*0201B2MHIV Gag (SLYNTVATL) Tetramer Protein (HLH-HP2H6) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ile 308 (HLA-A*0201) Ile 21 - Met 119 (B2M) SLYNTVATL peptide (Accession # AAA59606.1 (HLA-A*0201) P61769-1 (B2M) SLYNTVATL).
Catalog No. 50-253-4723
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.