Learn More
Hanna Pharmaceutical Supply Co., Inc. Paclitaxel Set, Vented, INTERLINK Injection Site, 0.2 Micron Filter, 10 drops/mL, 107""
Supplier:  Hanna Pharmaceutical Supply Co., Inc. 2C7557
Encompass_Preferred
Paclitaxel Set Vented Spike with Polyethylene Lined Tubing and INTERLINK Injection Site, 0.2 Micron Filter, Male Luer Lock Adapter. Approximately 10 drops/mL. Approximate length 107" (2.7 m). Non-DEHP Pump Segment. Sterile Fluid Path. Fluid path does not contain DEHP; outer layer of polyethylene lined tubing contains DEHP. 48/CS
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.
Catalog No. NC1844055
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.