Learn More
ABclonal Technology PE Rabbit anti- CD162/PSGL-1
SDP
Supplier:  ABclonal Technology A25949100T
Encompass_Preferred
Involved in leukocyte adhesive activation Acts upstream of or within leukocyte tethering or rolling Predicted to be located in plasma membrane raft and uropod Predicted to be active in plasma membrane Is expressed in brain and thymus primordium Human ortholog(s) of this gene implicated in carotid artery disease Orthologous to human SELPLG (selectin P ligand)
Catalog No. 50-000-12601
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.