Learn More
LifeSensors PE-SUMOSTAR KAN 20UG
SDP
Supplier:  LifeSensors PE11010020
Encompass_Preferred
The pE-SUMOstar vector is for the expression of recombinant SUMOstar fusion proteins (containing an N-terminal His6 tag) in E. coli. Production is driven by T7 RNA polymerase-promoter system, with kanamycin resistance facilitating plasmid selection and stability.
Catalog No. 50-103-7428
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.