Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEA15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517326
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517326 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517326 Supplier Invitrogen™ Supplier No. PA517326
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

PEA15 is a death effector domain (DED)-containing protein predominantly expressed in the central nervous system, particularly in astrocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEA15
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene PEA15
Gene Accession No. Q15121, Q5U318, Q62048
Gene Alias 15 kDa phosphoprotein enriched in astrocytes; astrocytic phosphoprotein PEA-15; HMAT1; homolog of mouse MAT-1 oncogene; HUMMAT1H; mammary transforming gene 1; mammary transforming gene 1, mouse, homolog of; MAT1; MAT1H; OTTHUMP00000029887; Pea15; PEA-15; Pea15a; PED; phosphoprotein enriched in astrocytes 15; phosphoprotein enriched in astrocytes 15A; phosphoprotein enriched in diabetes; Pkcs15; proliferation and apoptosis adaptor protein 15
Gene Symbols PEA15, Pea15a
Host Species Rabbit
Immunogen Synthetic peptide corresponding residue surrounding Leu60 of human PEA-15.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18611, 364052, 8682
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.