Learn More
Invitrogen™ PEAK1 Polyclonal Antibody

Description
Antibody detects endogenous levels of total SGK269.
Specifications
Specifications
| Antigen | PEAK1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | PEAK1 |
| Gene Accession No. | Q69Z38, Q9H792 |
| Gene Alias | 1110049L02Rik; 9530046P14; AA690169; BE132841; C230081A13Rik; Inactive tyrosine-protein kinase PEAK1; KIAA2002; mKIAA2002; NKF3 kinase family member; Peak1; pseudopodium enriched atypical kinase 1; pseudopodium-enriched atypical kinase 1; Sgk269; sugen kinase 269; Tyrosine-protein kinase SgK269 |
| Gene Symbols | PEAK1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.