Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Peanut oil

Catalog No. p-178789
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC416855000 500 mL Glass bottle
AC416850025 2.5 L Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC416855000 Supplier Thermo Scientific Chemicals Supplier No. 416855000
Only null left
Add to Cart

CAS: 8002-03-7

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8002-03-7
MDL Number MFCD00131805
Synonym Arachis oil, Groundnut oil

Specifications

Color Yellow
Physical Form Viscous Liquid
Quantity 500 mL
Flash Point 283°C
Merck Index 14, 7059
Solubility Information Solubility in water: insoluble. Other solubilities: very slightly soluble in alcohol,miscible with light petroleum
Concentration or Composition (by Analyte or Components) Saturated fatty acids (chain length <C16): 0.4% max.
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 0.915
Density 0.9150g/mL
Chemical Name or Material Peanut oil
Show More Show Less
EINECSNumber 232-296-4
RTECSNumber RX2830000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.