Learn More
3M™ Peanut Protein Rapid Kit L25PNT, Sold by AquaPhoenix Scientific
Shop All AquaPhoenix Scientific Products

Description
- Results visible at 11 ± 1 min. after application of sample
- Rapid, qualitative immunochromatographic lateral flow test cartridges designed for detection of allergenic food proteins
- Designed to detect both processed and unprocessed allergen proteins.
- Test can accommodate environmental swab, CIP rinse water and food samples
Specifications
Specifications
| Format | Kit |
| Product Type | Peanut Protein Rapid Kit |
| Test Type | Allergen |
| For Use With (Application) | Screening peanut proteins in CIP final rinse water, environmental swab samples, food ingredients and processed food products |
| Quantity | 25/Cs. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.