Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEBP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 360700
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-070-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-070-0 Supplier Invitrogen™ Supplier No. 360700
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

PEBP1 Polyclonal Antibody for Western Blot, ICC/IF, IP, ELISA

PBP binds ATP, opioids and phosphatidylethanolamine, exhibiting a lower affinity for phosphatidylinositol and phosphatidylcholine. This serine protease inhibitor inhibits thrombin, neuropsin and chymotrypsin but not trypsin, tissue type plasminogen activator and elastase. PBP contains hippocampal cholinergic neurostimulating peptide (HCNP), which may be involved in the function of the presynaptic cholinergic neurons of the central nervous system. HCNP increases the production of choline acetyltransferase but not acetylcholinesterase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEBP1
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene PEBP1
Gene Accession No. P30086, P31044, P70296
Gene Alias 23 kDa morphine-binding protein; Basic cytosolic 21 kDa protein; epididymis luminal protein 210; epididymis secretory protein Li 34; epididymis secretory protein Li 96; HCNP; HCNPpp; HEL-210; HEL-S-34; HEL-S-96; Hippocampal cholinergic neurostimulating peptide; KIAA0299; LDGF; MDGF; MOCA; neuropolypeptide h3; P23K; PBP; Pbp1; Pbpr; PEBP; PEBP1; PEBP-1; phosphatidylethanolamine binding protein; phosphatidylethanolamine binding protein 1; phosphatidylethanolamine-binding protein; phosphatidylethanolamine-binding protein 1; PPBP; prostatic binding protein; Prostatic-binding protein; Raf kinase inhibitor protein; Raf kinase inhibitory protein; Raf-1 inhibitor protein; Raf-1 kinase inhibitor protein; RKIP; similar to phosphatidylethanolamine binding protein; zgc:56033; zgc:76942
Gene Symbols PEBP1
Host Species Rabbit
Immunogen Synthetic peptide derived from the N-terminal region of human RKIP (Raf kinase inhibitor protein).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23980, 29542, 5037
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.