Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEBP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519142
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519142 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519142 Supplier Invitrogen™ Supplier No. PA519142
Only null left
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.

The phosphatidylethanolamine -binding proteins, including PEBP4, are an evolutionarily conserved family of proteins with pivotal biologic functions, such as lipid binding and inhibition of serine proteases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEBP4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Pebp4
Gene Accession No. Q96S96
Gene Alias CORK1; CORK-1; cousin-of-RKIP 1 protein; epididymis secretory protein Li 300; GWTM1933; HEL-S-300; hPEBP4; MGC22776; PEBP4; PEBP-4; phosphatidylethanolamine binding protein 4; phosphatidylethanolamine-binding protein 4; PRO4408; Protein cousin-of-RKIP 1; UNQ1933/PRO4408
Gene Symbols Pebp4
Host Species Goat
Immunogen Synthetic peptide sequence (RERASEPKHKNQAE) corresponding to the C-terminus amino acids of PEBP4 (aa 210-224).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 157310
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.