Learn More
MyBioSource CD31 ELISA Kit
SDP
Supplier:  MyBioSource MBS74291596STRIPWELLS
Encompass_Preferred
The MyBioSource PECAM-1/CD31 ELISA Kit quantifies PECAM-1/CD31 in various biological fluids using a specific antibody-coated 96-well plate. Samples are added, followed by a biotinylated detection antibody, HRP-conjugated streptavidin, and TMB substrate. Color intensity after adding the stop solution, indicating PECAM-1/CD31 levels, is measured at 450 nm.
Catalog No. NC2736322
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.