Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PECI Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. p-7209318 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-2858 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-2858 Supplier Proteintech Group Inc Supplier No. 203831AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PECI is an auxiliary enzyme that catalyzes an isomerization step required for the beta-oxidation of unsaturated fatty acids.

Specifications

Antigen PECI
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene ECI2
Gene Accession No. O75521, Q9WUR2
Gene Alias ACBD2, D3,D2 enoyl CoA isomerase, DBI related protein 1, Dodecenoyl CoA isomerase, DRS 1, DRS1, HCA88, KIAA0536, PECI
Gene Symbols ECI2
Host Species Rabbit
Immunogen PECI Fusion Protein Ag14211
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10455, 23986
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.