Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Pectinase (A. niger)

Catalog No. 89956397 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-956-397 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-956-397 Supplier Abnova™ Supplier No. P5284

Used for Enzyme

#REF!

Specifications

For Use With (Application) Enzyme Assay
Name Pectinase (A. niger)
Quantity 250 mg
Storage Requirements Store at 4°C on dry atmosphere.
Regulatory Status RUO
Common Name PECTINASE
Assays Pectinase Assay; Plant protoplast isolation.
Biological Activity ≥20 units per mg dry weight. One Unit releases 1 umole of D-galacturonic acid from polygalacturonic acid per minute at 37°C, pH 5.0.
Species Fungi
Protein Tag Untagged
Expression System Native protein chromatographically purified from Aspergillus niger
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.