Learn More
Bioassay Systems QuantiChrom™ Pectinase Assay Kit. For quantitative determination of pectinase activity determination in biological samples.
SDP
Catalog No. 502255864 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of pectinase activity determination in biological samples. Key Features: Linear detection range from 9.2-100 U/L pectinase activity in a 96-well plate assay. Simple and convenient 40 minute "Add-Mix-Measure" type assay. High-throughput format and compatible with laboratory liquid handling systems. No heating required. The kit does not use toxic materials. Refer to the previously established DNS method. Method: OD600nm. Samples: Enzyme extracts, agriculture and biological samples. Species: Plant, fungal tissues, juice, bacteria, etc. Procedure: Assay takes 40 min. Kit size: 100 tests. Detection limit: 9.2 U/L.
This item is not returnable. View return policy
Catalog No. 50-225-5864 Supplier Bioassay Systems Supplier No. DPEC100
For quantitative determination of pectinase activity determination in biological samples. Key Features: Linear detection range from 9.2-100 U/L pectinase activity in a 96-well plate assay. Simple and convenient 40 minute "Add-Mix-Measure" type assay. High-throughput format and compatible with laboratory liquid handling systems. No heating required. The kit does not use toxic materials. Refer to the previously established DNS method. Method: OD600nm. Samples: Enzyme extracts, agriculture and biological samples. Species: Plant, fungal tissues, juice, bacteria, etc. Procedure: Assay takes 40 min. Kit size: 100 tests. Detection limit: 9.2 U/L.