Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PEDF Monoclonal Antibody (C3)
GREENER_CHOICE

Catalog No. p-200124246
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA541660 200 μL
PIMA543629 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA541660 Supplier Invitrogen™ Supplier No. MA541660
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

PEDF Monoclonal Antibody for Western Blot, IHC (P)

PEDF is a noninhibitory serpin with neurotrophic, anti-angiogenic, and anti-tumorigenic properties. It is a 50 kDa glycoprotein produced and secreted in many tissues throughout the body. A major component of the anti-angiogenic action of PEDF is the induction of apoptosis in proliferating endothelial cells. In addition, PEDF is able to inhibit the activity of angiogenic factors, such as VEGF and FGF-2. The neuroprotective effects of PEDF are achieved through suppression of neuronal apoptosis induced by peroxide, glutamate, or other neurotoxins. The recent identification of a lipase-linked cell membrane receptor for PEDF (PEDF-R) that binds to PEDF with high affinity (1) should facilitate further elucidation of the underlying mechanisms of this pluripotent serpin. To date, PEDF-R is the only signaling receptor known to be used by a serpin family member. The unique range of PEDF activities implicate it as a potential therapeutic agent for the treatment of vasculature-related neurodegenerative diseases, such as age-related macular degeneration (AMD) and proliferative diabetic retinopathy (PDR). PEDF also has the potential to be useful in the treatment of various angiogenesis-related diseases including a number of cancers.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PEDF
Applications Immunohistochemistry (Paraffin), Western Blot, Western Blot
Classification Monoclonal
Clone C3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene SERPINF1
Gene Accession No. P36955, P97298
Gene Alias AI195227; alpha-2 antiplasmin; caspin; Cell proliferation-inducing gene 35 protein; Dmrs91; EPC 1; EPC1; EPC-1; OI12; OI6; PEDF; Pedfl; PIG 35; PIG35; pigment epithelium derived factor; pigment epithelium-derived factor; Sdf3; SDF-3; serine (or cysteine) peptidase inhibitor, clade F, member 1; serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 1; serine (or cysteine) proteinase inhibitor, clade F), member 1; serine (or cysteine) proteinase inhibitor, clade F, member 1; serpin F1; serpin family F member 1; serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 1; serpin peptidase inhibitor, clade F, member 1; SERPINF 1; Serpinf1; serpin-F1; sserpin family F member 1; stromal cell derived factor 3; stromal cell-derived factor 3; testis tissue sperm-binding protein Li 70n
Gene Symbols SERPINF1
Host Species Mouse
Immunogen Recombinant protein Pigment Epithelium Derived Factor. The antigen corresponds to amino acid range 19-417 of the target protein.
Purification Method Protein A/G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20317, 287526, 5176
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.