Learn More
Marketlab Pedia-Poser Pediatric Immobilizer Chair
Description
- Helps minimize retakes, reducing radiation exposure and saving time and money.
- Adjustable back and head-tilt support.
- Polyethylene chair components are x-ray lucent for excellent x-ray contrast.
- Saddle horn supports a genital lead shield.
- Nonporous polyethylene is easy to clean.
- Lockable casters.
- Views: C-Spine (AP, Lateral, Oblique), Chest (AP, Lateral), Abdominal (AP), Magnified Airway (AP, Lateral), Standing Lower Extremities (AP).
Specifications
Specifications
| Product Type | Pediatric Immobilizer |
| For Use With (Equipment) | X-Ray Systems |