Learn More
Thermo Scientific™ PEEK Fingertight Fittings
GSA_VA
Catalog No. 03052434 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
One-Piece PEEK Fingertight Fitting
One-Piece PEEK Long Fingertight Fitting
PEEK Wing Nut w/PEEK Ferrule
Description:
1/16 in.
1/16 in., 0.37 in. Head
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Description
03-052-434 PEEK Wing Nut w/PEEK Ferrule 1/16 in.
03-052-421 One-Piece PEEK Fingertight Fitting 1/16 in., 0.37 in. Head
03-052-422 One-Piece PEEK Long Fingertight Fitting 1/16 in., 0.37 in. Head
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03-052-434 Supplier Thermo Scientific™ Supplier No. F300X
Add to Cart
Add to Cart
Ensure HPLC quality and robustness with Thermo Scientific™ PEEK Fingertight Fittings.
Ensure HPLC quality and robustness with Thermo Scientific™ PEEK Fingertight Fittings. Machined for reliability, these fittings resist cracking, breaking, thread stripping and leaking in both low- and high-pressure applications. Fingertight plumbing ensures ease of use. The fittings are biocompatible for use across a broad range of applications.
  • Resist cracking, breaking, thread stripping and leaking in both low and high pressure applications
  • Biocompatible for a broad range of applications
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Material PEEK
For Use With (Equipment) LC Tubing
Description 1/16 in.
Type PEEK Wing Nut w/PEEK Ferrule
Quantity 10 Pack
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.